Merck
  • Time since onset of disease and individual clinical markers associate with transcriptional changes in uncomplicated dengue.
Time since onset of disease and individual clinical markers associate with transcriptional changes in uncomplicated dengue.
PLoS neglected tropical diseases (2015-03-15)
Cornelia A M van de Weg, Henk-Jan van den Ham, Maarten A Bijl, Fatih Anfasa, Fatiha Zaaraoui-Boutahar, Beti E Dewi, Leonard Nainggolan, Wilfred F J van IJcken, Albert D M E Osterhaus, Byron E E Martina, Eric C M van Gorp, Arno C Andeweg
ZUSAMMENFASSUNG
Dengue virus (DENV) infection causes viral haemorrhagic fever that is characterized by extensive activation of the immune system. The aim of this study is to investigate the kinetics of the transcriptome signature changes during the course of disease and the association of genes in these signatures with clinical parameters. Sequential whole blood samples from DENV infected patients in Jakarta were profiled using affymetrix microarrays, which were analysed using principal component analysis, limma, gene set analysis, and weighted gene co-expression network analysis. We show that time since onset of disease, but not diagnosis, has a large impact on the blood transcriptome of patients with non-severe dengue. Clinical diagnosis (according to the WHO classification) does not associate with differential gene expression. Network analysis however, indicated that the clinical markers platelet count, fibrinogen, albumin, IV fluid distributed per day and liver enzymes SGOT and SGPT strongly correlate with gene modules that are enriched for genes involved in the immune response. Overall, we see a shift in the transcriptome from immunity and inflammation to repair and recovery during the course of a DENV infection. Time since onset of disease associates with the shift in transcriptome signatures from immunity and inflammation to cell cycle and repair mechanisms in patients with non-severe dengue. The strong association of time with blood transcriptome changes hampers both the discovery as well as the potential application of biomarkers in dengue. However, we identified gene expression modules that associate with key clinical parameters of dengue that reflect the systemic activity of disease during the course of infection. The expression level of these gene modules may support earlier detection of disease progression as well as clinical management of dengue.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~0.025 M in H2O