Merck
  • EGFR inhibitor Gefitinib attenuates posterior capsule opacification in vitro and in the ex vivo human capsular bag model.
EGFR inhibitor Gefitinib attenuates posterior capsule opacification in vitro and in the ex vivo human capsular bag model.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie (2014-12-05)
Christian Wertheimer, Jakob Siedlecki, Daniel Kook, Wolfgang J Mayer, Armin Wolf, Annemarie Klingenstein, Anselm Kampik, Kirsten Eibl-Lindner
ZUSAMMENFASSUNG
Posterior capsule opacification (PCO) occurs as a common complication after cataract surgery. Gefitinib is a selective inhibitor of the epidermal growth factor receptor (EGFR) which represents a potential pharmacological target for PCO prevention. In this in vitro study, we assessed the effect and biocompatibility of Gefitinib in PCO prophylaxis. The effect of Gefitinib on the key pathological features of PCO was assessed in vitro. We determined growth in the human capsular bag model, prepared from sixteen cadaver eyes that underwent sham cataract surgery. Furthermore, two lens epithelial cell lines, HLE-B3 and FHL-124, were used to determine concentration-based effects on cell proliferation. In addition, cell-migration, matrix-contraction, and cell spreading were investigated. To exclude toxic concentrations, Gefitinib was assessed for its biocompatibility on six different human ocular cell types from the anterior and posterior segment of the eye. Gefitinib significantly increased the time until confluence of the capsular bag compared to controls (p < 0.001)). In both human lens epithelial cell lines (HLE-B3 and FHL-124), proliferation decreased significantly and as equally strong after incubation with Gefitinib (p < 0.001), as did chemotactic migration (p = 0.004), matrix contraction (p = 0.001), and cell-spreading (p = 0.001). At the IC50 concentration, Gefitinib was well tolerated by six different human ocular cell types of the anterior and posterior segment. The specific EGFR inhibitor Gefitinib might become of clinical relevance in PCO prophylaxis as it attenuated cellular growth and other pathological PCO factors in the ex vivo human capsular bag model and in two human lens epithelial cell lines, while showing good biocompatibility in vitro.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Supelco
Dimethylsulfoxid, analytical standard