Merck
  • Combination of intrauterine growth restriction and a high-fat diet impairs cholesterol elimination in rats.
Combination of intrauterine growth restriction and a high-fat diet impairs cholesterol elimination in rats.
Pediatric research (2014-08-15)
Erin K Zinkhan, Jeanette R Chin, Jennifer M Zalla, Baifeng Yu, Ben Numpang, Xing Yu, Chengshe Jiang, Christopher W Callaway, Robert A McKnight, Lisa Joss-Moore, Robert H Lane
ZUSAMMENFASSUNG
Intrauterine growth restriction (IUGR) increases the risk of adult-onset hypercholesterolemia. High-fat diet (HFD) consumption potentiates IUGR-induced increased cholesterol. Cholesterol is converted to bile acids by Cyp7a1 in preparation for excretion. We hypothesized that IUGR rats fed a HFD will have increased cholesterol, decreased Cyp7a1 protein levels, and decreased bile acids compared to control rats fed a HFD. At day 21, IUGR and control pups were placed on one of three diets: a regular chow or one of two HFDs containing 1% or 2% cholesterol. Cholesterol levels and hepatic Cyp7a1 protein levels were quantified a postnatal week 28. Both HFDs increased serum cholesterol levels in control rats, and HFD fed IUGR rats had further increased serum cholesterol up to 35-fold. Both HFDs increased hepatic cholesterol levels, and IUGR further increased hepatic cholesterol levels up to fivefold. IUGR decreased hepatic Cyp7a1 protein up to 75%, and hepatic bile acids up to 54%. IUGR increased cholesterol and bile acids and decreased Cyp7a1 protein in rats fed a HFD without changing food intake. These findings suggest that IUGR increases the vulnerability of HFD fed rats to hypercholesterolemia via decreased cholesterol conversion to bile acids.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Propylenglycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Propylenglykol, meets USP testing specifications
Supelco
Propylenglykol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,2-Propandiol, ACS reagent, ≥99.5%
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
USP
Propylenglycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,2-Propandiol, ReagentPlus®, 99%
Sigma-Aldrich
Stickstoff, ≥99.998%
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
1,2-Propandiol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
1,2-Propandiol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Supelco
1,2-Propandiol, analytical standard