Merck
  • Autoimmunity as a candidate for the etiopathogenesis of Meniere's disease: detection of autoimmune reactions and diagnostic biomarker candidate.
Autoimmunity as a candidate for the etiopathogenesis of Meniere's disease: detection of autoimmune reactions and diagnostic biomarker candidate.
PloS one (2014-10-21)
Sung Huhn Kim, Jin Young Kim, Hyun Jin Lee, Mia Gi, Bo Gyung Kim, Jae Young Choi
ZUSAMMENFASSUNG
Meniere's disease is an inner ear disorder that can manifest as fluctuating vertigo, sensorineural hearing loss, tinnitus, and aural fullness. However, the pathologic mechanism of Meniere's disease is still unclear. In this study, we evaluated autoimmunity as a potential cause of Meniere's disease. In addition we tried to find useful biomarker candidates for diagnosis. We investigated the protein composition of human inner ear fluid using liquid column mass spectrometry, the autoimmune reaction between circulating autoantibodies in patient serum and multiple antigens using the Protoarray system, the immune reaction between patient serum and mouse inner ear tissues using western blot analysis. Nine proteins, including immunoglobulin and its variants and interferon regulatory factor 7, were found only in the inner ear fluid of patients with Meniere's disease. Enhanced immune reactions with 18 candidate antigens were detected in patients with Meniere's disease in Protoarray analysis; levels of 8 of these antigens were more than 10-fold higher in patients than in controls. Antigen-antibody reactions between mouse inner ear proteins with molecular weights of 23-48 kDa and 63-75 kDa and patient sera were detected in 8 patients. These findings suggest that autoimmunity could be one of the pathologic mechanisms behind Meniere's disease. Multiple autoantibodies and antigens may be involved in the autoimmune reaction. Specific antigens that caused immune reactions with patient's serum in Protoarray analysis can be candidates for the diagnostic biomarkers of Meniere's disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
SAFC
Natriumchlorid -Lösung, 5 M
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Glycin
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Glycin, Pharmaceutical Secondary Standard; Certified Reference Material