Merck
  • Altered oxidative status and integrin expression in cyclosporine A-treated oral epithelial cells.
Altered oxidative status and integrin expression in cyclosporine A-treated oral epithelial cells.
Toxicology mechanisms and methods (2014-11-25)
Ahmadreza Sardarian, Azadeh Andisheh Tadbir, Fatemeh Zal, Fatemeh Amini, Aida Jafarian, Fatemeh Khademi, Zohreh Mostafavi-Pour
ZUSAMMENFASSUNG
Cyclosporine A (CsA) is an immunosuppressive agent administered to transplant patients. A well-known reported oral side effect of CsA consumption is gingival overgrowth (GO). Changes in the expression of integrins occurring in the gingiva following CsA treatment have been reported but these reports are mainly concerned with the connective tissue of the gingiva. In this study we targeted the alterations in the oral epithelium using KB cells, an oral epithelial cell line. Cultured oral epithelial cells were treated with increasing concentrations of CsA (0.1, 1 and 10 µg/mL) and the molecular changes involving antioxidant enzymes [glutathione peroxidase (GPx) and glutathione reductase (GR)] and the level of reactive oxygen species (ROS) were measured. Quantitative real-time PCR was used to assess the expression of selected integrins (α2, α5 and β1). At CsA concentration above 0.1 µg/mL GPx demonstrated an increase in activity while GR activity and the level of reduced glutathione were diminished (p < 0.05). α5 and β1 integrin were downregulated at all treatment concentrations of CsA while α2 integrin presented this effect at concentrations above 1 µg/mL (p < 0.05). The results suggest a possible role for oxidative stress and the altered expression of integrins in the pathology of CsA-induced gingival overgrowth.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Natriumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumphosphat, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Natriumphosphat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
tert-Butylhydroperoxid -Lösung, 70 wt. % in H2O
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
tert-Butylhydroperoxid -Lösung, 5.0-6.0 M in decane