Merck
Interpenetrating polymer network hydrogel scaffolds for artificial cornea periphery.
Journal of materials science. Materials in medicine (2015-02-11)
Rachel Parke-Houben, Courtney H Fox, Luo Luo Zheng, Dale J Waters, Jennifer R Cochran, Christopher N Ta, Curtis W Frank
ZUSAMMENFASSUNG
Three-dimensional scaffolds based on inverted colloidal crystals (ICCs) were fabricated from sequentially polymerized interpenetrating polymer network (IPN) hydrogels of poly(ethyleneglycol) and poly(acrylic acid). This high-strength, high-water-content IPN hydrogel may be suitable for use in an artificial cornea application. Development of a highly porous, biointegrable region at the periphery of the artificial cornea device is critical to long-term retention of the implant. The ICC fabrication technique produced scaffolds with well-controlled, tunable pore and channel dimensions. When surface functionalized with extracellular matrix proteins, corneal fibroblasts were successfully cultured on IPN hydrogel scaffolds, demonstrating the feasibility of these gels as materials for the artificial cornea porous periphery. Porous hydrogels with and without cells were visualized non-invasively in the hydrated state using variable-pressure scanning electron microscopy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
1,2-Dichlorethan, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Butanon, ACS reagent, ≥99.0%
Sigma-Aldrich
Wasser, Deionized
Sigma-Aldrich
N-Hydroxysuccinimid, 98%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Acrylsäure, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
1,2-Dichlorethan, anhydrous, 99.8%
Supelco
Wasser
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Osmiumtetroxid, ReagentPlus®, 99.8%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Osmiumtetroxid -Lösung, 4 wt. % in H2O
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free