Merck
  • Serum autoantibodies in pristane induced lupus are regulated by neutrophil gelatinase associated lipocalin.
Serum autoantibodies in pristane induced lupus are regulated by neutrophil gelatinase associated lipocalin.
Clinical immunology (Orlando, Fla.) (2014-06-28)
Rahul D Pawar, Beatrice Goilav, Yumin Xia, Haoyang Zhuang, Leal Herlitz, Westley H Reeves, Chaim Putterman
ZUSAMMENFASSUNG
The onset of autoantibodies in systemic autoimmunity can be the result of a breakdown in tolerance at multiple checkpoints. Genetic, hormonal, and immunological factors can combine with environmental influences to accelerate the onset of disease and aggravate disease outcome. Here, we describe a novel mechanism relating to the regulatory role of Neutrophil Gelatinase Associated Lipocalin (NGAL) in modulating the levels of autoantibodies in pristane induced lupus. Following a single injection of pristane intraperitoneally, NGAL expression was induced in both the serum and spleen. Furthermore, NGAL deficient mice were more susceptible to the induction of pristane stimulated autoimmunity, and displayed higher numbers of autoantibody secreting cells and increased expression of activation induced cytidine deaminase (AID) and other inflammatory mediators in the spleen. In contrast, kidney damage was milder in NGAL deficient mice, indicating that NGAL was detrimental in autoantibody mediated kidney disease. These studies indicate that NGAL plays differential roles in different tissues in the context of lupus, and suggest a previously unrecognized role for NGAL in adaptive immunity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
Pristan, synthetic, ≥98% (GC)
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Phosphatase-Substrat, 5 mg tablets
Sigma-Aldrich
4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, tablet
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Sigma-Aldrich
4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Phosphatase-Substrat, powder
Sigma-Aldrich
4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, tablet
Sigma-Aldrich
Phosphatase-Substrat, 40 mg tablets
Sigma-Aldrich
4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, tablet
Sigma-Aldrich
Phosphatase-Substrat, 100 mg capsules
SAFC
Pristan, synthetic, liquid, sterile-filtered, BioReagent
Sigma-Aldrich
Phosphatase-Substrat, 40 mg capsules
Sigma-Aldrich
Phosphatasesubstrat, Suitable for manufacturing of diagnostic kits and reagents
Sigma-Aldrich
Mouse MCP-1 / CCL2 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat MCP-1 / CCL2 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat MCP-1 / CCL2 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Mouse MCP-1 / CCL2 ELISA Kit, for cell and tissue lysates