Merck
  • Analgesic activity of Eugenia jambolana leave constituent: a dikaempferol rhamnopyranoside from ethyl acetate soluble fraction.
Analgesic activity of Eugenia jambolana leave constituent: a dikaempferol rhamnopyranoside from ethyl acetate soluble fraction.
Pharmaceutical biology (2014-07-16)
Madhu Cholenahalli Lingaraju, Shikha Anand, Venkanna Balaganur, Rashmi Rekha Kumari, Amar Sunil More, Dinesh Kumar, Brijesh Kumar Bhadoria, Surendra Kumar Tandan
ZUSAMMENFASSUNG
Eugenia jambolana Lam. (Myrtaceae) is a medicinal plant used in folk medicine for the treatment of diabetes, inflammation, and pain. We investigated the antinociceptive effect of kaempferol-7-O-α-l-rhamnopyranoside]- 4'-O-4'-[kaempferol-7-O-α-l-rhamnopyranoside (EJ-01), isolated from the E. jambolana leaves. EJ-01 (3, 10, and 30 mg kg(-1), orally) was assessed for peripheral (formalin-nociception and acetic acid-writhing) and central (hot plate and tail flick test) analgesic activity in mice and the in vitro anti-inflammatory activity (25, 50, and 100 µg mL(-1)) in lipopolysaccharide (LPS)-stimulated RAW 264.7 cells. EJ-01 (10 and 30 mg kg(-1)) significantly inhibited mean writhing counts (37.74 and 36.83) in acetic acid writhing and paw licking time (55.16 and 45.66 s) in the late phase of the formalin test as compared with the respective control (60.66 and 104.33 s). EJ-01 did not show analgesic activity in central pain models. Significant reduction in the tumor necrosis factor (TNF)-α (295.48, 51.20, and 49.47 pg mL(-1)) and interleukin (IL)-1β (59.38, 20.08, and 15.46 pg mL(-1)) levels were observed in EJ-01-treated medium (25, 50, and 100 µg mL(-1)) as compared with vehicle-treated control values (788.67 and 161.77 pg mL(-1)), respectively. Significant reduction in total nitrite plus nitrate (NOx) levels (70.80 nmol) was observed in the EJ-01-treated medium (100 µg mL(-1)) as compared with the vehicle-treated value (110.41 nmol). EJ-01 is a valuable analgesic constituent of E. jambolana leaves and this study supports the pharmacological basis for the use of this plant in traditional medicine for curing inflammatory pain.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Sulfanilamid, ≥98%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Formaldehyd -Lösung, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Formaldehyd -Lösung, tested according to Ph. Eur.
Supelco
Sulfanilamid Schmelzpunktstandard, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dexamethason, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sulfanilamid, puriss. p.a., ≥98% (calc. to the dried substance)
Supelco
Dimethylsulfoxid, analytical standard
Sigma-Aldrich
Dexamethason, meets USP testing specifications
USP
Sulfanilamid, United States Pharmacopeia (USP) Reference Standard
USP
Sulfanilamid, United States Pharmacopeia (USP) Reference Standard