Merck
  • A chromatin modifier regulates Sertoli cell response to mono-(2-ethylhexyl) phthalate (MEHP) via tissue inhibitor of metalloproteinase 2 (TIMP2) signaling.
A chromatin modifier regulates Sertoli cell response to mono-(2-ethylhexyl) phthalate (MEHP) via tissue inhibitor of metalloproteinase 2 (TIMP2) signaling.
Biochimica et biophysica acta (2014-08-26)
Chong Zhang, Jun-Hao Lai, Bin Hu, Shun Zhang, Jie Zhao, Wei Li
ZUSAMMENFASSUNG
Epigenetic silencing mechanisms are essential for regulating germ cell apoptosis in response to different stimuli during complicated spermatogenesis. Herein, we report the potential signaling events related to up-regulation of metastasis associated protein 1 (Mta1), a master chromatin modifier, during mono-(2-ethylhexyl) phthalate (MEHP)-induced Sertoli cells (SCs) injury. Mta1 up-regulation correlated to the gradual increases of MYC expression in MEHP-treated SCs. Selective knockdown of MYC abolished MEHP-induced activation of Mta1, suggesting that MYC may regulate the Mta1 signaling following MEHP injury. Furthermore, MTA1 acted as a specific corepressor of tissue inhibitor of metalloproteinase 2 (Timp2) during SCs injury. Mta1 repressed Timp2 expression either directly by recruiting histone deacetylase 2 onto the Timp2 promoter or indirectly by enhancing NF-κB-mediated inflammatory responses during MEHP injury. This transcriptional and post-translational down-regulation of Timp2/TIMP2 expression consequently resulted in the stimulated activation of matrix metalloproteinase 2 (MMP2) in SCs, which should ultimately promote germ cell death upon MEHP insult. From a functional standpoint, inhibition of endogenous Mta1 expression along with anti-inflammation treatment in cultured SCs could rescue MEHP-inhibited TIMP2 and subsequently rebalanced MMP2 activity to the control level. Together with the recently reported essential role of TIMP2/MMP2 signaling in MEHP-induced specific disruption of junctional complexes in the seminiferous epithelium, our results further substantiate a critical role of Mta1 in the control of SCs response to MEHP stimulation. The MYC/Mta1/TIMP2 circuit may serve as an important scavenger mechanism to help to maintain the capacity of damaged SCs to support germ cell development following MEHP injury.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Deoxycholsäure Natriumsalz, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Deoxycholsäure Natriumsalz, ≥97% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
SAFC
Deoxycholsäure Natriumsalz
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Selensäure, 98%
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Linolsäure, technical, 58-74% (GC)