Merck
  • Demyelination causes adult CNS progenitors to revert to an immature state and express immune cues that support their migration.
Demyelination causes adult CNS progenitors to revert to an immature state and express immune cues that support their migration.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2015-01-09)
Sarah Moyon, Anne Laure Dubessy, Marie Stephane Aigrot, Matthew Trotter, Jeffrey K Huang, Luce Dauphinot, Marie Claude Potier, Christophe Kerninon, Stephane Melik Parsadaniantz, Robin J M Franklin, Catherine Lubetzki
ZUSAMMENFASSUNG
The declining efficiency of myelin regeneration in individuals with multiple sclerosis has stimulated a search for ways by which it might be therapeutically enhanced. Here we have used gene expression profiling on purified murine oligodendrocyte progenitor cells (OPCs), the remyelinating cells of the adult CNS, to obtain a comprehensive picture of how they become activated after demyelination and how this enables them to contribute to remyelination. We find that adult OPCs have a transcriptome more similar to that of oligodendrocytes than to neonatal OPCs, but revert to a neonatal-like transcriptome when activated. Part of the activation response involves increased expression of two genes of the innate immune system, IL1β and CCL2, which enhance the mobilization of OPCs. Our results add a new dimension to the role of the innate immune system in CNS regeneration, revealing how OPCs themselves contribute to the postinjury inflammatory milieu by producing cytokines that directly enhance their repopulation of areas of demyelination and hence their ability to contribute to remyelination.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Zitronensäure, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Saccharose, ≥99.5% (GC)