Merck
In situ observation of radicals and molecular products during lignin pyrolysis.
ChemSusChem (2014-07-22)
Christian Bährle, Victoria Custodis, Gunnar Jeschke, Jeroen A van Bokhoven, Frédéric Vogel
ZUSAMMENFASSUNG
Lignin pyrolysis is a promising method for the sustainable production of phenolic compounds from biomass. However, detailed knowledge about the radicals involved in this process and their influence on the molecular products is missing. Herein, we report on the pyrolysis of hard- and softwood Klason lignins under inert gas conditions in a temperature range between 350-550 °C. During the pyrolysis process, the formed radicals were detected by in situ high-temperature electron paramagnetic resonance spectroscopy. The overall formation of volatile products during lignin pyrolysis was determined using thermogravimetric analysis. The volatile molecular products were characterized and quantified using GC-MS analysis. Major differences were observed between hardwood and softwood lignins. Hardwood lignins form more radicals and volatile products than softwood lignins at temperatures from 350 to 450 °C. In the late stages of the pyrolysis process at 550 °C radical quenching reactions become dominant in hardwood lignins. We identified the disproportionation of two semiquinone radicals to quinone and hydroquinone species as the most likely quenching reaction. Our results show that both the pyrolysis temperature and the type of lignin source have a major influence on radical formation and the molecular products during the depolymerization of lignin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Toluol, analytical standard
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis