Merck
  • Lysine deacetylases regulate the heat shock response including the age-associated impairment of HSF1.
Lysine deacetylases regulate the heat shock response including the age-associated impairment of HSF1.
Journal of molecular biology (2015-02-18)
Elena Zelin, Brian C Freeman
ZUSAMMENFASSUNG
Heat shock factor 1 (HSF1) is critical for defending cells from both acute and chronic stresses. In aging cells, the DNA binding activity of HSF1 deteriorates correlating with the onset of pathological events including neurodegeneration and heart disease. We find that DNA binding by HSF1 is controlled by lysine deacetylases with HDAC7, HDAC9, and SIRT1 distinctly increasing the magnitude and length of a heat shock response (HSR). In contrast, HDAC1 inhibits HSF1 in a deacetylase-independent manner. In aging cells, the levels of HDAC1 are elevated and the HSR is impaired, yet reduction of HDAC1 in aged cells restores the HSR. Our results provide a mechanistic basis for the age-associated regulation of the HSR. Besides HSF1, the deacetylases differentially modulate the activities of unrelated DNA binding proteins. Taken together, our data further support the model that lysine deacetylases are selective regulators of DNA binding proteins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
MISSION® shRNA, Purified DNA
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
USP
Dexamethason, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Mechlorethamin -hydrochlorid, 98%
Sigma-Aldrich
Monoklonaler ANTI-FLAG®-M1-Antikörper in Maus hergestellte Antikörper, clone M1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Dexamethason, meets USP testing specifications
Dexamethason, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethason, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethason, tested according to Ph. Eur.
Dexamethason, British Pharmacopoeia (BP) Assay Standard
Dexamethason für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Dexamethason für die Systemeignung, European Pharmacopoeia (EP) Reference Standard