Merck
  • Cephalosporinases associated with outer membrane vesicles released by Bacteroides spp. protect gut pathogens and commensals against β-lactam antibiotics.
Cephalosporinases associated with outer membrane vesicles released by Bacteroides spp. protect gut pathogens and commensals against β-lactam antibiotics.
The Journal of antimicrobial chemotherapy (2014-11-30)
Régis Stentz, Nikki Horn, Kathryn Cross, Louise Salt, Charles Brearley, David M Livermore, Simon R Carding
ZUSAMMENFASSUNG
To identify β-lactamase genes in gut commensal Bacteroides species and to assess the impact of these enzymes, when carried by outer membrane vesicles (OMVs), in protecting enteric pathogens and commensals. A deletion mutant of the putative class A β-lactamase gene (locus tag BT_4507) found in the genome of the human commensal Bacteroides thetaiotaomicron was constructed and a phenotypic analysis performed. A phylogenetic tree was built from an alignment of nine Bacteroides cephalosporinase protein sequences, using the maximum likelihood method. The rate of cefotaxime degradation after incubation with OMVs produced by different Bacteroides species was quantified using a disc susceptibility test. The resistance of Salmonella Typhimurium and Bifidobacterium breve to cefotaxime in liquid culture in the presence of B. thetaiotaomicron OMVs was evaluated by measuring bacterial growth. The B. thetaiotaomicron BT_4507 gene encodes a β-lactamase related to the CepA cephalosporinase of Bacteroides fragilis. OMVs produced by B. thetaiotaomicron and several other Bacteroides species, except Bacteroides ovatus, carried surface-associated β-lactamases that could degrade cefotaxime. β-Lactamase-harbouring OMVs from B. thetaiotaomicron protected Salmonella Typhimurium and B. breve from an otherwise lethal dose of cefotaxime. The production of membrane vesicles carrying surface-associated β-lactamases by Bacteroides species, which constitute a major part of the human colonic microbiota, may protect commensal bacteria and enteric pathogens, such as Salmonella Typhimurium, against β-lactam antibiotics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
PIPES, ≥99% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Dimethylsulfid, ≥99%, FCC, FG
Sigma-Aldrich
PIPES, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Dimethylsulfid, ≥99%
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
meso-2,3-Dimercaptobernsteinsäure, ~98%
Sigma-Aldrich
Dibenzothiophen, 98%
Sigma-Aldrich
Dimethylsulfid, anhydrous, ≥99.0%
Sigma-Aldrich
PIPES, BioXtra, ≥99% (titration)
Sigma-Aldrich
PIPES, BioXtra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Dibenzothiophen, ≥99%
Sigma-Aldrich
Dimethylsulfid, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Supelco
Dimethylsulfid, analytical standard
Supelco
Cefpodoxim, VETRANAL®, analytical standard
Supelco
Dibenzothiophen, analytical standard
Sigma-Aldrich
PIPES, anhydrous, free-flowing, Redi-Dri, ≥99%