Merck
Complement deficiencies limit CD20 monoclonal antibody treatment efficacy in CLL.
Leukemia (2014-05-03)
O Middleton, E Cosimo, E Dobbin, A M McCaig, C Clarke, A M Brant, M T Leach, A M Michie, H Wheadon
ZUSAMMENFASSUNG
Monoclonal antibodies (MAbs) form a central part of chronic lymphocytic leukaemia (CLL) treatment. We therefore evaluated whether complement defects in CLL patients reduced the induction of complement-dependent cytotoxicity (CDC) by using anti-CD20 MAbs rituximab (RTX) and ofatumumab (OFA). Ofatumumab elicited higher CDC levels than RTX in all CLL samples examined, particularly in poor prognosis cohorts (11q- and 17p-). Serum sample analyses revealed that 38.1% of patients were deficient in one or more complement components, correlating with reduced CDC responses. Although a proportion of patients with deficient complement levels initially induced high levels of CDC, on secondary challenge CDC activity in sera was significantly reduced, compared with that in normal human serum (NHS; P<0.01; n=52). In addition, a high CLL cell number contributed to rapid complement exhaustion. Supplementing CLL serum with NHS or individual complement components, particularly C2, restored CDC on secondary challenge to NHS levels (P<0.0001; n=9). In vivo studies revealed that complement components were exhausted in CLL patient sera post RTX treatment, correlating with an inability to elicit CDC. Supplementing MAb treatment with fresh-frozen plasma may therefore maintain CDC levels in CLL patients with a complement deficiency or high white blood cell count. This study has important implications for CLL patients receiving anti-CD20 MAb therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
SAFC
L-Glutamin
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Phenylessigsäure, 99%
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Sigma-Aldrich
Phenylessigsäure, ≥99%, FCC, FG
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Natriumpyruvat, BioXtra, ≥99%
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumpyruvat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland