Merck
  • Sepsis progression to multiple organ dysfunction in carotid chemo/baro-denervated rats treated with lipopolysaccharide.
Sepsis progression to multiple organ dysfunction in carotid chemo/baro-denervated rats treated with lipopolysaccharide.
Journal of neuroimmunology (2015-01-18)
Gino Nardocci, Aldo Martin, Sebastián Abarzúa, Jorge Rodríguez, Felipe Simon, Edison P Reyes, Claudio Acuña-Castillo, Cristina Navarro, Paula P Cortes, Ricardo Fernández
ZUSAMMENFASSUNG
Sepsis progresses to multiple organ dysfunction (MOD) due to the uncontrolled release of inflammatory mediators. Carotid chemo/baro-receptors could play a protective role during sepsis. In anesthetized male rats, we measured cardiorespiratory variables and plasma TNF-α, glucocorticoids, epinephrine, and MOD marker levels 90min after lipopolysaccharide (LPS) administration in control (SHAM surgery) and bilateral carotid chemo/baro-denervated (BCN) rats. BCN prior to LPS blunted the tachypneic response and enhanced tachycardia and hypotension. BCN-LPS rats also showed blunted plasma glucocorticoid responses, boosted epinephrine and TNF-α responses, and earlier MOD onset with a lower survival time compared with SHAM-LPS rats. Consequently, the complete absence of carotid chemo/baro-sensory function modified the neural, endocrine and inflammatory responses to sepsis. Thus, carotid chemo/baro-receptors play a protective role in sepsis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Sigma-Aldrich
Corticosteron, ≥98.5% (HPLC)
Sigma-Aldrich
Corticosteron, ≥92%
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O127:B8, purified by phenol extraction
Supelco
Hydrocortison, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Hydrocortison, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortison, meets USP testing specifications
Hydrocortison für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Hydrocortison, European Pharmacopoeia (EP) Reference Standard
Supelco
Corticosteron, VETRANAL®, analytical standard
Hydrocortison, British Pharmacopoeia (BP) Assay Standard