Merck
A detailed analysis of partial molecular volumes in DPPC/cholesterol binary bilayers.
Biochimica et biophysica acta (2014-08-26)
Tsubasa Miyoshi, Max Lönnfors, J Peter Slotte, Satoru Kato
ZUSAMMENFASSUNG
We examined the volumetric behavior of the dipalmitoylphosphatidylcholine (DPPC)/cholesterol binary bilayer system with high accuracy and more cholesterol concentrations to reveal the detailed molecular states in the liquid-disordered (Ld) phase, the liquid-ordered (Lo) phase and the gel phase. We measured the average specific volume of the binary bilayer at several temperatures by the neutral flotation method and calculated the average volume per molecule to estimate the partial molecular volumes of DPPC and cholesterol in each phase. As a result, we found that the region with intermediate cholesterol concentrations showed a more complicated behavior than expected from simple coexistence of Ld and Lo domains. We also measured fluorescence decay of trans-parinaric acid (tPA) added into the binary bilayer with more cholesterol concentrations to get further insight into the cholesterol-induced formation of the Lo phase. On the basis of these results we discuss the molecular interaction between DPPC and cholesterol molecule in the Lo phase and the manner of Ld/Lo phase coexistence.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Deuteriumoxid, filtered, 99.8 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.8 atom % D
Sigma-Aldrich
1,2-Dipalmitoyl-sn-Glycero-3-Phosphocholin aus Staphylococcus aureus, ≥99% (TLC)
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
1,2-Dipalmitoyl-sn-Glycero-3-Phosphocholin aus Staphylococcus aureus, semisynthetic, ≥99%
Sigma-Aldrich
Deuteriumoxid, 70 atom % D
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.
Sigma-Aldrich
Deuteriumoxid, 60 atom % D
Sigma-Aldrich
Deuteriumoxid, 99.9 atom % D