Merck
  • Expansion of human mesenchymal stromal cells from fresh bone marrow in a 3D scaffold-based system under direct perfusion.
Expansion of human mesenchymal stromal cells from fresh bone marrow in a 3D scaffold-based system under direct perfusion.
PloS one (2014-07-16)
Adam Papadimitropoulos, Elia Piccinini, Sophie Brachat, Alessandra Braccini, David Wendt, Andrea Barbero, Carsten Jacobi, Ivan Martin
ZUSAMMENFASSUNG
Mesenchymal stromal/stem cell (MSC) expansion in conventional monolayer culture on plastic dishes (2D) leads to progressive loss of functionality and thus challenges fundamental studies on the physiology of skeletal progenitors, as well as translational applications for cellular therapy and molecular medicine. Here we demonstrate that 2D MSC expansion can be entirely bypassed by culturing freshly isolated bone marrow nucleated cells within 3D porous scaffolds in a perfusion-based bioreactor system. The 3D-perfusion system generated a stromal tissue that could be enzymatically treated to yield CD45- MSC. As compared to 2D-expanded MSC (control), those derived from 3D-perfusion culture after the same time (3 weeks) or a similar extent of proliferation (7-8 doublings) better maintained their progenitor properties, as assessed by a 4.3-fold higher clonogenicity and the superior differentiation capacity towards all typical mesenchymal lineages. Transcriptomic analysis of MSC from 5 donors validated the robustness of the process and indicated a reduced inter-donor variability and a significant upregulation of multipotency-related gene clusters following 3D-perfusion--as compared to 2D-expansion. Interestingly, the differences in functionality and transcriptomics between MSC expanded in 2D or under 3D-perfusion were only partially captured by cytofluorimetric analysis using conventional surface markers. The described system offers a multidisciplinary approach to study how factors of a 3D engineered niche regulate MSC function and, by streamlining conventional labor-intensive processes, is prone to automation and scalability within closed bioreactor systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
Selen, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Trypanblau, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Selen, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, suitable for microscopy
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trypanblau, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
USP
Dexamethason, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Selen, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Dexamethason, meets USP testing specifications
Sigma-Aldrich
Selen, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Natriumpyruvat, BioXtra, ≥99%
Sigma-Aldrich
Natriumpyruvat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethason, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethason, VETRANAL®, analytical standard