Merck
Endogenous bone regeneration is dependent upon a dynamic oxygen event.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (2014-04-23)
Mimi C Sammarco, Jennifer Simkin, Danielle Fassler, Alex J Cammack, Aaron Wilson, Keith Van Meter, Ken Muneoka
ZUSAMMENFASSUNG
Amputation of the digit tip within the terminal phalangeal bone of rodents, monkeys, and humans results in near-perfect regeneration of bone and surrounding tissues; however, amputations at a more proximal level fail to produce the same regenerative result. Digit regeneration is a coordinated, multifaceted process that incorporates signaling from bioactive growth factors both in the tissue matrix and from several different cell populations. To elucidate the mechanisms involved in bone regeneration we developed a novel multi-tissue slice-culture model that regenerates bone ex vivo via direct ossification. Our study provides an integrated multi-tissue system for bone and digit regeneration and allows us to circumvent experimental limitations that exist in vivo. We used this slice-culture model to evaluate the influence of oxygen on regenerating bone. Micro-computed tomography (µCT) and histological analysis revealed that the regenerative response of the digit is facilitated in part by a dynamic oxygen event, in which mutually exclusive high and low oxygen microenvironments exist and vacillate in a coordinated fashion during regeneration. Areas of increased oxygen are initially seen in the marrow and then surrounding areas of vasculature in the regenerating digit. Major hypoxic events are seen at 7 days postamputation (DPA 7) in the marrow and again at DPA 12 in the blastema, and manipulation of oxygen tensions during these hypoxic phases can shift the dynamics of digit regeneration. Oxygen increased to 21% oxygen tension can either accelerate or attenuate bone mineralization in a stage-specific manner in the regenerative timeline. These studies not only reveal a circumscribed frame of oxygen influence during bone regeneration, but also suggest that oxygen may be one of the primary signaling influences during regeneration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Eosin Y -Lösung, 5 wt. % in H2O
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y Dinatriumsalz, certified by the BSC
Sigma-Aldrich
Eosin Y Dinatriumsalz, ≥85 % by HPLC
Sigma-Aldrich
Eosin Y, 75% (HPLC)