Merck
Effect of extrusion on phytochemical profiles in milled fractions of black rice.
Food chemistry (2015-02-24)
Huihui Ti, Ruifen Zhang, Mingwei Zhang, Zhencheng Wei, Jianwei Chi, Yuanyuan Deng, Yan Zhang
ZUSAMMENFASSUNG
The phytochemical profile and antioxidant activities of unprocessed and extruded milled fractions of black rice were investigated. Extrusion increased the free phenolics, anthocyanins and oxygen radical absorbance capacity (ORAC) and decreased the bound forms. The total phenolics, anthocyanins and ORAC increased by 12.6%, 5.4% and 19.7%, respectively, in bran. Extrusion decreased both free and bound phenolics and anthocyanins while ORAC values decreased by 46.5%, 88.4% and 33.1%, respectively, in polished rice and by 71.2%, 87.9% and 14.7%, respectively, in brown rice. A total of seven phenolics, gallic, chlorogenic, vanillic, caffeic, syringic, p-coumaric and ferulic acids, were detected in both forms. Cyanidin 3-glucoside (Cy-3-G), cyanidin 3-rutinoside and peonidin 3-glucoside were also detected with Cy-3-G found in the highest amounts in unprocessed and extruded rice bran. These results provide the basis for the development of different milled fractions of extruded black rice with balanced nutritional characteristics for today's functional food markets.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Kaliumchlorid, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥98%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Natriumacetat, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%