Merck
  • Supplementation with Pfaffia glomerata (Sprengel) Pedersen does not affect androgenic-anabolic parameters in male rats.
Supplementation with Pfaffia glomerata (Sprengel) Pedersen does not affect androgenic-anabolic parameters in male rats.
Journal of ethnopharmacology (2014-12-17)
Ney Felipe Fernandes, Anderson Joel Martino-Andrade, Ana Carolina Dos Santos Lourenço, Juliane Centeno Muller, Katherinne Maria Spercoski, Fabiola Nihi, Marilis Dallarmi Miguel, Vinícius Bednarczuk de Oliveira, Paulo Roberto Dalsenter, Rosana Nogueira Morais
ZUSAMMENFASSUNG
Paffia spp (Amaranthacea) has a widespread use of in Brazil as a possible hormonal supplement and a substitute of Panax ginseng, although information on its reproductive effects is missing. To evaluated possible anabolic-androgenic or anti-androgenic effects of Pfaffia glomerata (PG) extract using intact eight-months-old male rats and pre-pubertal castrated rats. Three different dose levels of PG (8.5, 30 and 85 mg/kg/day) were administered to eight-months-old rats for 28 days or to castrated males for 7 days (Hershberger assay). In the experiment with intact animals, 24h fecal samples were collected for quantification of fecal metabolites of androgens throughout treatment. At the end of the treatment period, animals were euthanized for evaluation of serum testosterone, reproductive organ weights, number of spermatids per testis, diameter of seminiferous tubules and cross-sectional area of soleus muscle fibers. In the Hershberber assay, androgenic or anti-androgenic effects were evaluated by the weights of androgen-dependent tissues: ventral prostate, seminal vesicle, glans penis and levator ani muscle/bulbocavernosus muscle. No effects were observed in the concentrations of fecal metabolites of androgens monitored during the treatment of intact eight-months-old rats. Moreover, at the end of treatment, no changes were seen in any of the investigated parameters. In the Hershberger assay, the PG extract did not induce androgenic or anti-androgenic effects at the dose levels tested. Significant effects were only observed in animals treated with testosterone and testosterone plus flutamide, which were used as positive controls for androgenicity and anti-androgenicity, respectively. At the dose levels tested, PG extract does not induce anabolic-androgenic or anti-androgenic effects in rats.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Ethanol
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Testosteronpropionat, tested according to Ph. Eur.