Merck
  • SIRT1 protects against apoptosis by promoting autophagy in degenerative human disc nucleus pulposus cells.
SIRT1 protects against apoptosis by promoting autophagy in degenerative human disc nucleus pulposus cells.
Scientific reports (2014-12-17)
Wei Jiang, Xuemei Zhang, Jie Hao, Jieliang Shen, Ji Fang, Wen Dong, Dawu Wang, Xiaojun Zhang, Wei Shui, Yi Luo, Liangbo Lin, Quanhe Qiu, Bin Liu, Zhenming Hu
ZUSAMMENFASSUNG
SIRT1 could protect degenerative human NP cells against apoptosis, and there were extensive and intimate connection between apoptosis and autophagy. Up to now, the role of autophagy in the process of human IVD degeneration is unclear. We sought to explore the relationship between autophagy and human IVD degeneration and to understand whether autophagy is involved in the protective effect of SIRT1 against apoptosis in NP cells. Our results showed that the autophagosomes number, the mRNA level of LC3 and Beclin-1, the protein expression of LC3-II/I and Beclin-1, decreased in NP from DDD. Resveratrol could increase the protein expression of LC3-II/I and Beclin-1, and reduce apoptosis in degenerative NP cells. In contrast, the protein levels of LC3-II/I and Beclin-1 were down-regulated and apoptosis level was significantly up-regulated in treatment with nicotinamide or SIRT1-siRNA transfection. Further analysis identified that the expression of cleaved Caspase3 and apoptosis incidence significantly increased with the pretreatment of bafilomycin A, whether resveratrol was added or not. These suggested that autophagy may play an important role in IVD degeneration, and SIRT1 protected degenerative human NP cells against apoptosis via promoting autophagy. These findings would aid in the development of novel therapeutic approaches for degenerative disc disease treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Nicotinamid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Bafilomycin A1 aus Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Nicotinamid, ≥99.5% (HPLC)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Supelco
Niacinamid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nicotinamid, ≥98% (HPLC), powder
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥95% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥99%
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, Grade AA-1
USP
Niacinamid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid, pkg of 10 mg (per vial)
Sigma-Aldrich
Niacinamid, meets USP testing specifications
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid Hydrat, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotid, pkg of 50 mg (per vial)
Supelco
Dimethylsulfoxid, analytical standard
Sigma-Aldrich
2,2′-Bichinolin-4,4′-dicarbonsäure Dinatriumsalz Dihydrat, ≥98% (HPLC)
Supelco
Resveratrol, analytical standard