Merck
  • Detection of cystatin C biomarker for clinical measurement of renal disease by developed ELISA diagnostic kits.
Detection of cystatin C biomarker for clinical measurement of renal disease by developed ELISA diagnostic kits.
Journal of translational medicine (2014-09-14)
Renren Jiang, Chao Xu, Xiaoli Zhou, Tianhao Wang, Gang Yao
ZUSAMMENFASSUNG
Human cystatin C (HCC) is a potential biomarker for tubular damage and impaired renal function. It is difficult to obtain efficient paired monoclonal antibodies against HCC with low molecular to meet the requirements for clinical application The present study was to establish a stable and repeatable measurement for HCC with self-made monoclonal antibodies (McAbs) and Variable domain of heavy chain of heavy-chain antibody (VHHs) increase the sensitivity. With hybridoma technology and phage display technology: R-HCC as a screening antigen and N-HCC as the detector for antigens to obtain the specific antibody and established an enzyme-linked immunosorbent assay for human cystatin C using self-made McAbs and VHHs. We have successfully obtained three McAbs; 5 F2, 4E4, 1E11 and four VHHs; 3-2, 3-24, 3-33 and 4-5 which were specific for HCC. The measurement of HCC was established with the self-made monoclonal antibodies and VHHs with a high sensitivity the lower limit of detection at 0.5 ng/ml and the detection range at 0.5 ~ 31.3 ng/ml. Our data provides a new approach for paired antibody screening and testing of the small molecular biomarker with a single dominant epitope, with the important biological and clinical significance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Thymidin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Hypoxanthin, ≥99.0%
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Thymidin, ≥99%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Hypoxanthin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
L-Lysin -monohydrochlorid, reagent grade, ≥98% (HPLC)
Supelco
L-Lysin -monohydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Thymidin, ≥99.0% (HPLC)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Aminopterin, powder
Sigma-Aldrich
L-Lysin -monohydrochlorid, BioUltra, ≥99.5% (AT)
Lysin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aminopterin, powder, BioReagent, suitable for cell culture
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aminopterin, Hybri-Max, 50 ×, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Supelco
L-Lysin-hydrochlorid -Lösung, 100 mM amino acid in 0.1 M HCl, analytical standard
Supelco
L-Lysin -monohydrochlorid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Didanosin Unreinheit A,, European Pharmacopoeia (EP) Reference Standard