Merck
  • Cholesterol-overloaded HDL particles are independently associated with progression of carotid atherosclerosis in a cardiovascular disease-free population: a community-based cohort study.
Cholesterol-overloaded HDL particles are independently associated with progression of carotid atherosclerosis in a cardiovascular disease-free population: a community-based cohort study.
Journal of the American College of Cardiology (2015-01-31)
Yue Qi, Jie Fan, Jing Liu, Wei Wang, Miao Wang, Jiayi Sun, Jun Liu, Wuxiang Xie, Fan Zhao, Yan Li, Dong Zhao
ZUSAMMENFASSUNG
Cholesterol-overloaded high-density lipoprotein (HDL) particles exert a negative impact on the antiatherogenic function of HDL in experimental studies. However, it remains unclear whether cholesterol-overloaded HDL particle is involved in the development of atherosclerosis in humans. The objective of this study was to explore whether cholesterol-overloaded HDL particles are associated with the progression of carotid atherosclerosis in a cardiovascular disease-free population. Baseline HDL particle number was measured using nuclear magnetic resonance spectroscopy in 930 participants ages 45 to 74 years in a community-based cohort study. An estimate of cholesterol molecules per HDL particle (HDL-C/P ratio) was calculated as the ratio of HDL cholesterol to HDL particles. HDL-C/P ratio was categorized as <41.0 (lowest), 41.0 to 46.9, 47.0 to 52.9, and ≥53.0 (highest) using a fixed increment method. Modified Poisson regression was used to assess the association between HDL-C/P ratio and 5-year progression of carotid atherosclerosis as indicated by progression of carotid plaques and change in total plaque area (TPA). Mean baseline HDL-C/P ratio was 46.4 ± 9.3 (range 23.8 to 86.9). Baseline HDL-C/P ratio was significantly associated with 5-year progression of carotid atherosclerosis. Participants with the highest HDL-C/P ratio had 1.56-fold (95% confidence interval: 1.14 to 2.13; p = 0.006) increased progression compared with those with the lowest level. Among participants without baseline plaque, TPA in re-examination was larger by 9.4 mm(2) in the subgroup with the highest level when compared with the lowest level. Our findings suggest that cholesterol-overloaded HDL particles are independently associated with the progression of carotid atherosclerosis. This may explain why in recent trials raising HDL cholesterol was not beneficial. This study strongly suggests that the combination of cholesterol content and particle number determines the antiatherogenic function of HDLs, rather than either parameter alone.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.