Merck
  • A biodegradable antibiotic-impregnated scaffold to prevent osteomyelitis in a contaminated in vivo bone defect model.
A biodegradable antibiotic-impregnated scaffold to prevent osteomyelitis in a contaminated in vivo bone defect model.
European cells & materials (2014-06-09)
J S McLaren, L J White, H C Cox, W Ashraf, C V Rahman, G W Blunn, A E Goodship, R A Quirk, K M Shakesheff, R Bayston, B E Scammell
ZUSAMMENFASSUNG
Open fractures are at risk of serious infection and, if infected, require several surgical interventions and courses of systemic antibiotics. We investigated a new injectable formulation that simultaneously hardens in vivo to form a porous scaffold for bone repair and delivers antibiotics at high concentrations to the local site of infection. Duration of antimicrobial activity against Staphylococcus aureus was determined using the serial plate transfer test. Ultimate compressive strength and porosity of the material was measured with and without antibiotics. The material was evaluated in vivo in an ovine medial femoral condyle defect model contaminated with S. aureus. Sheep were sacrificed at either 2 or 13 weeks and the defect and surrounding bone assessed using micro-computed tomography and histology. Antimicrobial activity in vitro persisted for 19-21 days. Sheep with antibiotic-free material and bacteria became infected, while those with antibiotic-containing material and bacteria did not. Similarly, new bone growth was seen in uninoculated animals with plain polymer, and in those with antibiotic polymer with bacteria, but not in sheep with plain polymer and bacteria. The antibiotic-impregnated scaffolds were effective in preventing S. aureus infections whilst supporting bone growth and repair. If translated into clinical practice, this approach might reduce the need for systemic antibiotics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Clindamycin -hydrochlorid, lincosamide antibiotic