Merck
  • Antimicrobial properties of liposomal azithromycin for Pseudomonas infections in cystic fibrosis patients.
Antimicrobial properties of liposomal azithromycin for Pseudomonas infections in cystic fibrosis patients.
The Journal of antimicrobial chemotherapy (2014-11-25)
Venkata Saran Solleti, Moayad Alhariri, Majed Halwani, Abdelwahab Omri
ZUSAMMENFASSUNG
This work was carried out to construct a novel liposomal azithromycin formulation and examine its antimicrobial effects against Pseudomonas aeruginosa. The liposomal azithromycin formulation was prepared by the dehydration-rehydration vesicle method and its characterizations were tested. The MIC and the MBC of the liposomal formulation were determined by the microbroth dilution method. Liposomal azithromycin activity against biofilm-forming P. aeruginosa was assessed using a Calgary biofilm device. The effect of subinhibitory concentrations of liposomal azithromycin on bacterial virulence factors and motility studies was tested on P. aeruginosa strains. The bacteria and liposome interactions were studied using flow cytometry analysis. The toxicities of the liposomal formulation on erythrocytes and A549 lung cells were evaluated in vitro. The average diameter of the liposomal azithromycin was 406.07 ± 45 nm and the encapsulation efficiency was 23.8% ± 0.2%. The MIC and MBC values of liposomal azithromycin were significantly lower than those of free azithromycin. The liposomal azithromycin significantly reduced the bacteria in the biofilm and attenuated the production of different virulence factors; it also reduced the different patterns of bacterial motilities. By flow cytometry analysis data, it was shown that there are interactions of liposomes with the bacterial membranes. No significant haemolysis or cell toxicity was observed with the liposomal formulation. The results of this research indicate that this novel liposomal azithromycin formulation could be a useful therapy to enhance the safety and efficacy of azithromycin against P. aeruginosa-infected persons.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Trypanblau, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, suitable for microscopy
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Trypanblau, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Azithromycin
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Calcium Standard für AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
USP
Azithromycin, United States Pharmacopeia (USP) Reference Standard
Supelco
Calcium Ionenlösung für ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Azithromycin für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Azithromycin für die Preakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Azithromycin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.
USP
Azithromycin-Identität, United States Pharmacopeia (USP) Reference Standard