Merck
  • Active matrix metalloproteases are expressed early on and are high during the Barrett's esophagus malignancy sequence.
Active matrix metalloproteases are expressed early on and are high during the Barrett's esophagus malignancy sequence.
Scandinavian journal of gastroenterology (2015-01-07)
Akueni L Davelaar, Daniëlle Straub, Navtej S Buttar, Paul Fockens, Kausilia K Krishnadath
ZUSAMMENFASSUNG
Molecular processes underlying Barrett's malignant development are poorly understood. Matrix metalloproteases (MMPs) are enzymes involved in inflammation, tissue remodeling, and malignant development. Therefore, active MMPs may have a role in early metaplasia development and Barrett's esophagus' malignant progression. We desired to gain more insight into the role of MMPs during the Barrett's esophagus pathogenesis sequence. In a surgical Barrett's mouse model, and in nonmalignant Barrett's and malignant esophageal cell lines, the activity of MMPs was investigated using a MMP activatable probe. MMP activity was further validated in Barrett's esophagus and esophageal adenocarcinoma patient biopsies and was further differentiated by investigating MMP9 and MMP13 expressions. The mouse model showed probe activation in stromal cells early on in the esophagitis and metaplasia stages. MMP probe activation was higher in the Barrett's and cancer cell lines and biopsies as compared to normal cells and tissues. Co-immunostainings confirmed that, at the tissue level, the probe activation was mostly confined to CD45-positive stromal cells. MMP13 expression was highest in Barrett's metaplasia, whereas MMP9 was highest in the esophageal adenocarcinomas. During the Barrett's pathogenesis process, MMP activity is increased early on in the inflamed esophagus and remains high in metaplasia and esophageal adenocarcinoma. However, there is a switch of MMP13 to MMP9 expression once neoplasia develops. In the future, detecting specific MMP subtypes could be used for distinguishing nonmalignant from neoplastic Barrett's esophagus.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Glutamin
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis