Merck
  • Stimulation of electroporation-induced inward currents in glioblastoma cell lines by the heat shock protein inhibitor AUY922.
Stimulation of electroporation-induced inward currents in glioblastoma cell lines by the heat shock protein inhibitor AUY922.
Clinical and experimental pharmacology & physiology (2014-06-10)
Nai-Jung Chiang, Sheng-Nan Wu, Ching-An Kao, Yan-Ming Huang, Li-Tzong Chen
ZUSAMMENFASSUNG
Membrane electroporation (MEP) increases the electrical conductivity of the plasma membrane by addition of an external electrical field. Combining MEP-induced current (IMEP ) with antineoplastic agents has been increasingly considered as a new therapeutic manoeuvre, especially in the treatment of malignant gliomas. Thus, the aim of the present study was to evaluate the effect of AUY922 (AUY), a potent inhibitor of heat-shock protein 90 (HSP90), on IMEP in glioblastoma cells. The IMEP in glioblastoma cells (U373) was generated by repetitive hyperpolarization from -80 to -200 mV. The amplitude of IMEP was increased by AUY in a concentration-dependent manner, with an EC50 of 0.32 μmol/L. In addition AUY shortened the latency to IMEP generation. Before depolarization to +50 mV, hyperpolarization to -200 mV for 50 msec produced Ca(2+) influx and subsequently increased the amplitude of the Ca(2+) -activated K(+) current (IK(Ca) ). The amplitude of IK(Ca) and Ca(2+) influx was further increased by AUY through its ability to activate IMEP . Other HSP90 inhibitors, namely 17-(allylamino)-17-demethoxygeldanamycin (17-AAG; 1 μmol/L) and 6-chloro-9-[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]-9H-purin-2-amine (BIIB021; 1 μmol/L), only slightly (albeit significantly) increased the amplitude of IMEP in glioblastoma cells. A 50 msec depolarizing step elevated Ca(2+) influx and subsequently increased the amplitude of IK(Ca) in the presence of these three inhibitors. These data indicate that the AUY-mediated stimulation of IMEP and IK(Ca) in glioblastoma cells is independent of HSP90 inhibition. Moreover, these results indicate that AUY-stimulated IMEP and the subsequent activation of IK(Ca) may create important signalling events in glioblastoma cells. Thus, AUY is a drug that could potentially be used to augment the effectiveness of electrochemotherapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
SAFC
L-Glutamin
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Apamin, from bee venom, ≥95% (HPLC)
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nimodipin
Sigma-Aldrich
Apamin, synthetic, ≥97% (HPLC)
Sigma-Aldrich
17-(Allylamino)-17-demethoxygeldanamycin, ≥98% (HPLC), solid
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Nimodipin, Pharmaceutical Secondary Standard; Certified Reference Material
Nimodipin, European Pharmacopoeia (EP) Reference Standard