Merck
  • Seeding of mesenchymal stem cells into inner part of interconnected porous biodegradable scaffold by a new method with a filter paper.
Seeding of mesenchymal stem cells into inner part of interconnected porous biodegradable scaffold by a new method with a filter paper.
Dental materials journal (2015-03-10)
Katsuyuki Yamanaka, Katsushi Yamamoto, Yuhiro Sakai, Youko Suda, Yusuke Shigemitsu, Tadashi Kaneko, Koichi Kato, Tomohiro Kumagai, Yukio Kato
ZUSAMMENFASSUNG
An appropriate physical support provided by scaffolds creates a supportive environment that directs proliferation and differentiation of stem cells. However, it is difficult to homogenously inoculate stem cells into the inner part of scaffolds at high cell densities. In this study, mesenchymal stem cells were seeded into a hydroxyapatite/poly (D, L-lactic-co-glycolic acid) (HAP/PLGA) scaffold that had enough mechanical strength and porous 3-D structure. With an aid of a filter paper placed under the bottom of a HAP/PLGA block, the cells suspended in a culture medium flowed from the top to the bottom through interconnected pores in the scaffold, and distributed almost homogenously, as compared to cell distribution near the surface of the block by the conventional method using centrifugation or reduced pressure. This simple method with a filter paper may be useful in preparation of cell-scaffold complexes for tissue engineering.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,4-Dioxan, ACS reagent, ≥99.0%, contains ≤25 ppm BHT as stabilizer
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
1,4-Dioxan, suitable for HPLC, ≥99.5%
Sigma-Aldrich
1,4-Dioxan, ACS reagent, ≥99.0%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Supelco
1,4-Dioxan, analytical standard
Supelco
1,4-Dioxan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis