Merck
Bronchodilator, vasodilator and spasmolytic activities of Cymbopogon martinii.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society (2015-01-03)
K H Janbaz, A Qayyum, F Saqib, I Imran, M Zia-Ul-Haq, V de Feo
ZUSAMMENFASSUNG
Cymbopogon martinii (Cm.Cr) is traditionally used in south Asian communities for the management of multiple ailments including gastrointestinal, respiratory and vascular disorders and the present study was undertaken to validate these folkloric uses. The application of a methanol extract of the plant (Cm.Cr) to isolated rabbit jejunum preparation exhibited relaxation through decrease in magnitude and frequency of spontaneous contractions. The Cm.Cr also exerted relaxant effect on high K(+) (80 mM) induced contractions in isolated rabbit jejunum preparations. The Cm.Cr and its dichloromethane (Cm.Dcm) and aqueous (Cm.Aq) fractions also caused concentration-dependent relaxation in spontaneous and K(+) (80 mM) induced contractions which are comparables to effects produced by verapamil. Cm.Cr caused shifting of the Ca(2+)-curves toward right, suggesting the presence of a Ca(2+) channel blocking activity. Subsequently, Cm.Cr, Cm.Dcm and Cm.Aq caused relaxation of CCh (1 μM) and K(+) (80 mM) induced contractions in isolated rabbit tracheal preparations, suggesting that the observed relaxant effect can be mediated through antimuscarinic and/or Ca(2+) channel blocking activities. Cm.Cr tested against phenylephrine (PE; 1 μM) and K(+) (80 mM) induced contractions exhibited partial relaxation of isolated rabbit aortic preparations. The above-mentioned studies provided a scientific basis for the folkloric use of Cymbopogon martini in the management of multiple ailments in traditional systems of medicines.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumchlorid, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Magnesiumsulfat, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ammoniumhydroxid -Lösung, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Kaliumchlorid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Kaliumchlorid, Molecular Biology, ≥99.0%
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets