Merck
Structural changes of 6a-hydroxy-pterocarpans upon heating modulate their estrogenicity.
Journal of agricultural and food chemistry (2014-10-10)
Milou G M van de Schans, Jean-Paul Vincken, Toine F H Bovee, Alfredo David Cervantes, Madelon J Logtenberg, Harry Gruppen
ZUSAMMENFASSUNG
The isoflavonoid composition of an ethanolic extract of fungus-treated soybean sprouts was strongly altered by a combined acid/heat treatment. UHPLC-MS analysis showed that 6a-hydroxy-pterocarpans were completely converted to their respective, more stable, 6a,11a-pterocarpenes, whereas other isoflavonoids, from the isoflavone and coumestan subclasses, were affected to a much lesser extent (loss of ∼15%). Subsequently, mixtures enriched in prenylated 6a-hydroxy-pterocarpans (pools of glyceollin I/II/III and glyceollin IV/VI) or prenylated 6a,11a-pterocarpenes (pools of dehydroglyceollin I/II/III and dehydroglyceollin IV/VI) were purified, and tested for activity on both human estrogen receptors (ERα and ERβ). In particular, the response toward ERα changed, from agonistic for glyceollins to antagonistic for dehydroglyceollins. Toward ERβ a decrease in agonistic activity was observed. These results indicate that the introduction of a double bond with the concomitant loss of a hydroxyl group in 6a-hydroxy-pterocarpans extensively modulates their estrogenic activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Ammoniumsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Ammoniumsulfat, Molecular Biology, ≥99.0%
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%