Merck
  • Involvement of nitridergic and opioidergic pathways in the antinociception of gabapentin in the orofacial formalin test in mice.
Involvement of nitridergic and opioidergic pathways in the antinociception of gabapentin in the orofacial formalin test in mice.
Pharmacological reports : PR (2015-02-26)
Hugo F Miranda, Fernando Sierralta, Sebastian Lux, Rocío Troncoso, Natalia Ciudad, Ramiro Zepeda, Pilar Zanetta, Viviana Noriega, Juan Carlos Prieto
ZUSAMMENFASSUNG
Pain is one of the most common problems in clinical medicine. There is considerable evidence that pharmacologic approaches are the most widely used therapeutic options to ameliorate persistent or chronic pain. In this study it was evaluated the effect of l-NAME and naltrexone in the antinociception induced by administration of gabapentin in the orofacial formalin test of mice. The algesiometer assay was performed by the administration of 20 μl of 2% formalin solution injected into the upper right lip of each mouse. The dose of gabapentin that produces the 50% of the maximum possible effect (ED50) was significantly increased by the pretreatment with l-NAME or naltrexone. These results suggest that gabapentin produce antinociception partly via the activation nitridergic pathways and opioid system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Chlorwasserstoff – Methanol -Lösung, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Naltrexon -hydrochlorid
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%