Merck
  • The role of physico-chemical and bulk characteristics of co-spray dried L-leucine and polyvinylpyrrolidone on glidant and binder properties in interactive mixtures.
The role of physico-chemical and bulk characteristics of co-spray dried L-leucine and polyvinylpyrrolidone on glidant and binder properties in interactive mixtures.
International journal of pharmaceutics (2015-01-13)
Sharad Mangal, Felix Meiser, Satu Lakio, David Morton, Ian Larson
ZUSAMMENFASSUNG
In this study, polyvinylpyrrolidone (PVP) was spray dried with l-leucine (PVP-Leu) to create a prototype multifunctional interactive excipient. The physico-chemical and bulk properties such as particle size, surface composition, surface energy and bulk cohesion of PVP-Leu was measured and compared against pure spray dried PVP (PVP-SD). The mixing behaviour of these excipients and their effect on flow and binder activity of paracetamol was assessed. The mean particle sizes of PVP-Leu PVP-SD and PVP were 2.5, 2.1 and 21.9μm, respectively. Surface composition characterization indicated that l-leucine achieved higher concentrations on the surface compared to the bulk of the PVP-Leu particles. The surface energy of PVP-Leu was significantly lower compared to PVP-SD. In addition, PVP-Leu exhibited a significantly lower bulk cohesion compared PVP-SD. The excipients were blended with paracetamol and qualitative characterization indicated that PVP-Leu blended more homogeneously with paracetamol compared to PVP-SD. Both PVP-Leu and PVP-SD then exhibited a significantly improved binder activity compared to PVP. The flow of the paracetamol was markedly improved with PVP-Leu while PVP-SD and PVP had negligible effect on its flow. This study reveals how physico-chemical and bulk properties of such prototype interactive excipients can play a key role in determining multi-factorial excipient performance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
4-Acetamidophenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Laktose Monohydrat, ≥99% (HPLC), BioUltra
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
4-Acetamidophenol, BioXtra, ≥99.0%
Supelco
Laktose (Monohydrat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Acetamidophenol, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
D-Laktose Monohydrat, ≥98.0% (HPLC)
Sigma-Aldrich
4-Acetamidophenol, analytical standard
Sigma-Aldrich
Magnesiumstearat, technical grade
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-Laktose Monohydrat, ACS reagent
USP
Lactose Monohydrat, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Magnesiumstearat, puriss., meets analytical specification of Ph. Eur., BP, ≥90% stearic and palmitic acid basis, ≥40% stearic acid basis (GC), 4.0-5.0% Mg basis (calc on dry sub.)
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
D-Laktose Monohydrat, tested according to Ph. Eur.