Merck
  • Co-delivery of cisplatin and rapamycin for enhanced anticancer therapy through synergistic effects and microenvironment modulation.
Co-delivery of cisplatin and rapamycin for enhanced anticancer therapy through synergistic effects and microenvironment modulation.
ACS nano (2014-04-12)
Shutao Guo, C Michael Lin, Zhenghong Xu, Lei Miao, Yuhua Wang, Leaf Huang
ZUSAMMENFASSUNG
The tumor microenvironment plays an important role in the tumor's progression and metastasis. Therefore, successful alteration of this delicate setting against the tumor's favor can open a window for therapeutic efficacy. We have developed a modality to bring about treatment-induced alterations in the tumor microenvironment by employing the synergistic effects between two drugs. Co-delivery of rapamycin (RAPA), an mTOR inhibitor that may offer notable therapy through antiangiogenic activity, alongside cisplatin can foster significant potency as RAPA sensitizes A375 melanoma cells to cisplatin therapy through microenvironment modulation. However, encapsulation of these drugs into poly(lactic-co-glycolic acid) (PLGA) NPs was inefficient due to the incompatibility between the two free drugs and the polymer matrix. Here, we show cisplatin can be made hydrophobic by coating a nanoprecipitate (cores) of the drug with dioleoylphosphatidic acid (DOPA). These DOPA coated cisplatin cores are compatible with PLGA and can be coencapsulated in PLGA NPs alongside RAPA at a molar ratio to promote synergistic antitumor activity. The presence of the cisplatin cores significantly improved the encapsulation of RAPA into PLGA NPs. Furthermore, PLGA NPs containing both cisplatin cores and RAPA induced significant apoptosis on A375-luc human melanoma cells in vitro. Additionally, they inhibited the growth of A375-luc melanoma in a xenograft tumor model through modulation of the tumor vasculature and permitted enhanced penetration of NPs into the tumor.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cyclohexan, ACS reagent, ≥99%
Sigma-Aldrich
Cyclohexan, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N-Hydroxysuccinimid, 98%
Sigma-Aldrich
N,N-Diisopropylethylamin, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Diisopropylethylamin, 99.5%, biotech. grade
Sigma-Aldrich
3,4-Dihydroxy-L-Phenylalanin, ≥98% (TLC)
Sigma-Aldrich
Stearinsäure, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
N,N-Diisopropylethylamin, purified by redistillation, 99.5%
Sigma-Aldrich
1-Hexanol, reagent grade, 98%
Sigma-Aldrich
cis-Diamminplatin(II)-dichlorid, crystalline
Sigma-Aldrich
Stearinsäure, reagent grade, 95%
Sigma-Aldrich
Cyclohexan, anhydrous, 99.5%
Sigma-Aldrich
Stearinsäure, ≥95%, FCC, FG
Sigma-Aldrich
4-Methoxybenzoesäure, ReagentPlus®, 99%
Sigma-Aldrich
1-Hexanol, anhydrous, ≥99%
Supelco
Levodopa, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Stearinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cyclohexan, ACS reagent, ≥99%
Supelco
Stearinsäure, analytical standard
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diaminplatin(II)-dichlorid, ≥99.9% trace metals basis
Supelco
Stearinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Levodopa, United States Pharmacopeia (USP) Reference Standard
Supelco
Cyclohexan, analytical standard
Sigma-Aldrich
N-Hydroxysuccinimid, purum, ≥97.0% (T)
Sigma-Aldrich
N-Ethyldiisopropylamin, ≥98.0%
Sigma-Aldrich
p-Anissäure, ≥99%, FG
Stearinsäure, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hexylalkohol, natural, ≥98%, FCC, FG
Supelco
1-Hexanol, analytical standard