Merck
  • Biosynthesis of 15N-labeled cylindrospermopsin and its application as internal standard in stable isotope dilution analysis.
Biosynthesis of 15N-labeled cylindrospermopsin and its application as internal standard in stable isotope dilution analysis.
Analytical and bioanalytical chemistry (2014-07-30)
Katrin Kittler, Holger Hoffmann, Franziska Lindemann, Matthias Koch, Sascha Rohn, Ronald Maul
ZUSAMMENFASSUNG
Cylindrospermopsin (CYN) is a cyanobacterial toxin associated with human and animal poisonings. Due to its toxicity in combination with its widespread occurrence, the development of reliable methods for selective, sensitive detection and accurate quantification is mandatory. Liquid chromatography tandem mass spectrometry (LC-MS/MS) analysis using stable isotope dilution analysis (SIDA) represents an ideal tool for this purpose. U-[(15)N5]-CYN was synthesized by culturing Aphanizomenon flos-aquae in Na(15)NO3-containing cyanobacteria growth medium followed by a cleanup using graphitized carbon black columns and mass spectrometric characterization. Subsequently, a SIDA-LC-MS/MS method for the quantification of CYN in freshwater and Brassica matrices was developed showing satisfactory performance data. The recovery ranged between 98 and 103 %; the limit of quantification was 15 ng/L in freshwater and 50 μg/kg dry weight in Brassica samples. The novel SIDA was applied for CYN determination in real freshwater samples as well as in kale and in vegetable mustard exposed to toxin-containing irrigation water. Two of the freshwater samples taken from German lakes were found to be CYN-contaminated above limit of quantification (17.9 and 60.8 ng/L). CYN is systemically available to the examined vegetable species after exposure of the rootstock leading to CYN mass fractions in kale and vegetable mustard leaves of 15.0 μg/kg fresh weight and 23.9 μg/kg fresh weight, respectively. CYN measurements in both matrices are exemplary for the versatile applicability of the developed method in environmental analysis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥98%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Magnesiumsulfat, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumcarbonat, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Natriumcarbonat, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Zitronensäure, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumcarbonat, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Natriumcarbonat, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Magnesiumsulfat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Magnesiumsulfat, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)