Merck
  • HSP90 inhibition downregulates thymidylate synthase and sensitizes colorectal cancer cell lines to the effect of 5FU-based chemotherapy.
HSP90 inhibition downregulates thymidylate synthase and sensitizes colorectal cancer cell lines to the effect of 5FU-based chemotherapy.
Oncotarget (2014-10-10)
Ganji Purnachandra Nagaraju, Olatunji B Alese, Jerome Landry, Roberto Diaz, Bassel F El-Rayes
ZUSAMMENFASSUNG
Cell cycle progression and DNA synthesis are essential steps in cancer cell growth. Thymidylate synthase (TS) is a therapeutic target for 5FU. We tested the hypothesis that HSP90 transcriptional and functional inhibition can inhibit cell cycle progression, downregulate TS levels and sensitize colorectal cancer (CRC) cell lines to the effects of 5FU. Treatment with ganetespib (50 nM) for 24 hours inhibited cyclin D1 and pRb at the transcriptional and translational levels and induced p21, leading to G0/G1 cell cycle arrest in both CRC cell lines (HCT-116 and HT-29). This was associated with downregulation of E2F1 and its target gene TS. In addition, ganetespib inhibited PI3K/Akt and ERK signalling pathways. Similar effects were observed with HSP90 knockdown in both cell lines. Ganetespib sensitized CRC cell lines to the effects of oxaliplatin and 5FU. Similar effects were also observed in tumors from animals treated with ganetespib, oxaliplatin and 5FU. In this study, we present in vitro and animal data supporting that the targeting of HSP90 decreases CRC cell survival and proliferation. Ganetespib sensitizes CRC cell lines to the effects of 5FU-based chemotherapy. Combining HSP90 inhibitors with chemotherapy is a rational approach for future drug development in CRC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
2-Butanon, ACS reagent, ≥99.0%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Titan(IV)-chlorid, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
SAFC
L-Glutamin
Sigma-Aldrich
Titan(IV)-chlorid -Lösung, 1.0 M in methylene chloride
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Oxaliplatin, powder
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
USP
Natriumbicarbonat, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)