Merck
Involvement of dopamine signaling in the circadian modulation of interval timing.
The European journal of neuroscience (2014-04-03)
Ivana L Bussi, Gloria Levín, Diego A Golombek, Patricia V Agostino
ZUSAMMENFASSUNG
Duration discrimination within the seconds-to-minutes range, known as interval timing, involves the interaction of cortico-striatal circuits via dopaminergic-glutamatergic pathways. Besides interval timing, most (if not all) organisms exhibit circadian rhythms in physiological, metabolic and behavioral functions with periods close to 24 h. We have previously reported that both circadian disruption and desynchronization impaired interval timing in mice. In this work we studied the involvement of dopamine (DA) signaling in the interaction between circadian and interval timing. We report that daily injections of levodopa improved timing performance in the peak-interval procedure in C57BL/6 mice with circadian disruptions, suggesting that a daily increase of DA is necessary for an accurate performance in the timing task. Moreover, striatal DA levels measured by reverse-phase high-pressure liquid chromatography indicated a daily rhythm under light/dark conditions. This daily variation was affected by inducing circadian disruption under constant light (LL). We also demonstrated a daily oscillation in tyrosine hydroxylase levels, DA turnover (3,4-dihydroxyphenylacetic acid/DA levels), and both mRNA and protein levels of the circadian component Period2 (Per2) in the striatum and substantia nigra, two brain areas relevant for interval timing. None of these oscillations persisted under LL conditions. We suggest that the lack of DA rhythmicity in the striatum under LL - probably regulated by Per2 - could be responsible for impaired performance in the timing task. Our findings add further support to the notion that circadian and interval timing share some common processes, interacting at the level of the dopaminergic system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, Molecular Biology, ≥97.0%
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
E-64, protease inhibitor
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Natriumorthovanadat, ≥90% (titration)
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, ≥97.0%
Sigma-Aldrich
Natriumorthovanadat, 99.98% trace metals basis
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Supelco
Pefabloc® SC
Sigma-Aldrich
4-(2-Aminoethyl)benzolsulfonylfluorid -hydrochlorid, ≥97.0% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Saccharose, meets USP testing specifications
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)