Merck
  • Effect of aliskiren on circulating endothelial progenitor cells and vascular function in patients with type 2 diabetes and essential hypertension.
Effect of aliskiren on circulating endothelial progenitor cells and vascular function in patients with type 2 diabetes and essential hypertension.
American journal of hypertension (2014-07-06)
Athanasios E Raptis, Konstantinos P Markakis, Maria C Mazioti, Ignantios Ikonomidis, Eirini P Maratou, Dimitrios V Vlahakos, Eleni E Kotsifaki, Asterios N Voumvourakis, Alexandra G Tsirogianni, Vaia A Lambadiari, John P Lekakis, Sotirios A Raptis, George D Dimitriadis
ZUSAMMENFASSUNG
The aim of this study was to investigate the effects of aliskiren on vascular function and endothelial progenitor cells (EPCs) in patients with type 2 diabetes and essential hypertension. The study enrolled type 2 diabetic patients aged >50 years under stable glycemic control and first diagnosed mild essential hypertension. In phase A (n = 20), patients received aliskiren 150-300 mg daily for 3 months. In phase B (n = 12), hydrochlorothiazide (HCTZ) 12.5-25mg daily substituted for aliskiren for 3 more months. At baseline and at the end of each phase, we assessed (i) brachial blood pressure (BBP); (ii) central aortic systolic pressure (CSP), aortic augmentation index (Aix), and pulse wave velocity (PWV) as markers of arterial stiffness; (iii) brachial artery flow-mediated dilatation (FMD) as a marker of endothelial function; (iv) left ventricular (LV) twisting and untwisting as markers of LV function and (v) EPC numbers in culture of peripheral blood mononuclear cells. Aliskiren similarly reduced BBP and CSP, increased FMD (P < 0.001) and EPC numbers (P < 0.001), decreased PWV and Aix (P < 0.05), and improved LV twisting and untwisting (P < 0.05). Although substitution of HCTZ sustained BBP at similar levels, CSP and echocardiographic indices nearly returned at baseline levels, and the improvement of FMD, PWV, Aix, and EPC numbers was abolished. Aliskiren had a favorable effect on endothelial function and EPCs, reduced arterial stiffness, and improved LV twisting and untwisting. These effects were independent of BBP lowering, as they were not observed after the achievement of similar values of BBP with HCTZ.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)