Merck
Black phosphorus quantum dots.
Angewandte Chemie (International ed. in English) (2015-02-05)
Xiao Zhang, Haiming Xie, Zhengdong Liu, Chaoliang Tan, Zhimin Luo, Hai Li, Jiadan Lin, Liqun Sun, Wei Chen, Zhichuan Xu, Linghai Xie, Wei Huang, Hua Zhang
ZUSAMMENFASSUNG
As a unique two-dimensional nanomaterial, layered black phosphorus (BP) nanosheets have shown promising applications in electronics. Although mechanical exfoliation was successfully used to prepare BP nanosheets, it is still a challenge to produce novel BP nanostructures in high yield. A facile top-down approach for preparation of black phosphorus quantum dots (BPQDs) in solution is presented. The obtained BPQDs have a lateral size of 4.9±1.6 nm and thickness of 1.9±0.9 nm (ca. 4±2 layers). As a proof-of-concept application, by using BPQDs mixed with polyvinylpyrrolidone as the active layer, a flexible memory device was successfully fabricated that exhibits a nonvolatile rewritable memory effect with a high ON/OFF current ratio and good stability.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, ReagentPlus®, 99%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, anhydrous, 99.5%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, suitable for HPLC, ≥99%
Sigma-Aldrich
1-Methyl-2-pyrrolidon, biotech. grade, ≥99.7%
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichlormethan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tantal(V)-ethoxid, 99.98% trace metals basis
Supelco
Dichlormethan, analytical standard
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
1-Methyl-2-pyrrolidon, analytical standard
Sigma-Aldrich
Dichlormethan, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Dichlormethan, Selectophore, ≥99.5%
Tantal(V)-ethoxid, packaged for use in deposition systems