Merck
The effects of endothelial progenitor cells on rat atherosclerosis.
Biotechnology and applied biochemistry (2014-06-04)
Xianyou Wang, Feng Wang, Nana Li, Min Hu, Yun Chen, Mengqun Tan
ZUSAMMENFASSUNG
Atherosclerosis (AS) is a progressive disease characterized by endothelial injury and lipid aggregation in the arterial walls. Studies have reported that endothelial progenitor cells (EPCs) derived from the bone marrow (BM) might provide an endogenous repair mechanism by differentiating into endothelial cells to replace the dysfunctional endothelium. Our study aims to investigate the effect of EPCs derived from rat BM on AS. EPCs transduced by recombinant adeno-associated virus-green fluorescent protein (GFP) were transplanted into a rat AS model. After 2 months of transplantation, the localization of GFP-labeled cells, morphology, and lipid content in the aorta were examined. GFP-labeled EPCs were found in the endothelial monolayer of the artery vessel in the GFP/EPC group. Hematoxylin and eosin staining suggested that the lipid deposits in the aortic endothelium in the EPC/GFP group were less compared with those in the untreated group. Oil Red O staining of liver slices showed that lipid droplets were obviously decreased in the GFP/EPC group. The endothelial nitric oxide synthase and apolipoprotein E mRNA levels in the GFP/EPC group were significantly higher, but the intercellular cell adhesion molecule-1 mRNA level was significantly lower compared with the control group. The results suggest that EPCs derived from the BM can repair the injured endothelium and promote an atherosclerotic lesion regression. Therefore, EPCs may provide a useful tool for the treatment of AS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Propylenglycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Propylenglykol, meets USP testing specifications
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Propylenglykol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
1,2-Propandiol, ACS reagent, ≥99.5%
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Glycerin, FCC, FG
USP
Propylenglycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,2-Propandiol, ReagentPlus®, 99%
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
1,2-Propandiol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Supelco
Glycerin, analytical standard
Sigma-Aldrich
Glycerin, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
1,2-Propandiol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%