Merck
  • Desferrioxamine-caffeine (DFCAF) as a cell permeant moderator of the oxidative stress caused by iron overload.
Desferrioxamine-caffeine (DFCAF) as a cell permeant moderator of the oxidative stress caused by iron overload.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine (2014-10-01)
Elizabeth Carmen Pastrana Alta, Dibakar Goswami, M Teresa Machini, Daniel Menezes Silvestre, Cassiana Seimi Nomura, Breno Pannia Espósito
ZUSAMMENFASSUNG
Desferrioxamine (DFO) is a potent iron chelator used in the treatment of iron overload (IO) disorders. However, due to its low cell permeability and fast clearance, DFO administration is usually prolonged and of limited use for the treatment of IO in tissues such as the brain. Caffeine is a safe, rapidly absorbable molecule that can be linked to other compounds to improve their cell permeability. In this work, we successfully prepared and described DFO-caffeine, a conjugate with iron scavenging ability, antioxidant properties and enhanced permeation in the HeLa cell model.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Deferoxamin -mesylat (Salz), powder, ≥92.5% (TLC)
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Supelco
Trifluoressigsäure, analytical standard
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Calcein-AM -Lösung, 4 mM in DMSO, ≥90% (HPLC), solution
Deferoxamin -mesylat (Salz), European Pharmacopoeia (EP) Reference Standard