Merck
  • Simultaneous quantification by HPLC of the phenolic compounds for the crude drug of Prunus serotina subsp. capuli.
Simultaneous quantification by HPLC of the phenolic compounds for the crude drug of Prunus serotina subsp. capuli.
Pharmaceutical biology (2014-03-13)
Blanca Rivero-Cruz
ZUSAMMENFASSUNG
Prunus serotina Ehrenb. subsp. capuli (Cav.) McVaugh (Rosaceae), commonly known as "capulin", is a native North American tree, commercialized and used in folk medicine for the treatment of the hypertension, gastrointestinal illnesses, and cough. This work developed a suitable HPLC method for quantifying the major active constituents of the infusion of P. serotina, the most important preparation consumed by populations around the world. The analytical method was performed using a Fortis-RP column (150 mm × 4.6 mm; film thickness 5 µm). The mobile phase consisted of an isocratic acetate buffer solution (pH 2.7; A) and methanol (B) (65:35 v/v) at a flow rate of 1.0 mL min(-1). The proposed method was applied to the quantification of 1-3 in several samples of the leaves of P. serotina. The results indicated that amounts of 1-3 in the samples analyzed are uniform, and greater amounts of chlorogenic acid (2; 479.9 ± 33.6 µg g(-1), dry matter) along with hyperoside (1; 185.7 ± 55.3 µg g(-1), dry matter) were present. On the other hand, benzaldehyde (3; 118.2 ± 12.1 µg g(-1) dry matter) was found to be in lower concentration. A simple, sensitive, precise, and reproducible HPLC method for the simultaneous quantification of 1-3 in P. serotina was developed and validated. This is the first report on the quantification of 1-3 as active principles, and compound 1 was selected as a marker of P. serotina, which could be useful to guarantee the quality of the crude drug and herbal products.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumacetat, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natriumacetat, ACS reagent, ≥99.0%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Natriumacetat, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Chlorogensäure, ≥95% (titration)
Sigma-Aldrich
Benzaldehyd, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzaldehyd, purified by redistillation, ≥99.5%
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Supelco
Benzaldehyd, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumacetat, 99.995% trace metals basis
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Natriumacetat, >99%, FG
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Natriumacetat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumacetat, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Supelco
Essigsäure, analytical standard
Sigma-Aldrich
Natriumacetat -Lösung, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Benzaldehyd, ≥98%, FG, FCC
Sigma-Aldrich
Natriumacetat, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Benzaldehyd, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Natriumacetat, meets USP testing specifications, anhydrous
Supelco
Benzaldehyd, analytical standard
Sigma-Aldrich
Benzaldehyd, natural, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)