Merck
  • Human neural stem cell intracerebral grafts show spontaneous early neuronal differentiation after several weeks.
Human neural stem cell intracerebral grafts show spontaneous early neuronal differentiation after several weeks.
Biomaterials (2015-01-27)
Annette Tennstaedt, Markus Aswendt, Joanna Adamczak, Ursel Collienne, Marion Selt, Gabriele Schneider, Nadine Henn, Cordula Schaefer, Marie Lagouge, Dirk Wiedermann, Peter Kloppenburg, Mathias Hoehn
ZUSAMMENFASSUNG
Human neural stem cells (hNSCs) hold great promise for the treatment of neurological diseases. Considerable progress has been made to induce neural differentiation in the cell culture in vitro and upon transplantation in vivo [2] in order to explore restoration of damaged neuronal circuits. However, in vivo conventional strategies are limited to post mortem analysis. Here, we apply our developed first fate mapping platform to monitor neuronal differentiation in vivo by magnetic resonance imaging, bioluminescence imaging, and fluorescence imaging. Ferritin, Luciferase and GFP under neuronal-specific promoters for immature and mature neurons, respectively, were used to generate transgenic hNSCs. Differentiation-linked imaging reporter expression was validated in vitro. The time profile of spontaneous neuronal maturation after transplantation into mouse brain cortex demonstrated early neuronal differentiation within 6 weeks. Fully mature neurons expressing synaptogenesis were observed only after three months or longer. Our trimodal fate mapping strategy represents a unique non-invasive tool to monitor the time course of neuronal differentiation of transplanted stem cells in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
o-Xylen, reagent grade, ≥98.0%
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Kalium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
o-Xylen, anhydrous, 97%
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Kaliumhydrid, 30 wt % dispersion in mineral oil