Merck
  • Bidirectional amygdaloid control of neuropathic hypersensitivity mediated by descending serotonergic pathways acting on spinal 5-HT3 and 5-HT1A receptors.
Bidirectional amygdaloid control of neuropathic hypersensitivity mediated by descending serotonergic pathways acting on spinal 5-HT3 and 5-HT1A receptors.
Behavioural brain research (2015-01-06)
B Sagalajev, N Bourbia, E Beloushko, H Wei, A Pertovaara
ZUSAMMENFASSUNG
Amygdala is involved in processing of primary emotions and particularly its central nucleus (CeA) also in pain control. Here we studied mechanisms mediating the descending control of mechanical hypersensitivity by the CeA in rats with a peripheral neuropathy in the left hind limb. For drug administrations, the animals had a guide cannula in the right CeA and an intrathecal catheter or another guide cannula in the medullary raphe. Hypersensitivity was tested with monofilaments. Glutamate administration in the CeA produced a bidirectional effect on hypersensitivity that varied from an increase at a low-dose (9μg) to a reduction at high doses (30-100μg). The increase but not the reduction of hypersensitivity was prevented by blocking the amygdaloid NMDA receptor with a dose of MK-801 that alone had no effects. The glutamate-induced increase in hypersensitivity was reversed by blocking the spinal 5-HT3 receptor with ondansetron, whereas the reduction in hypersensitivity was reversed by blocking the spinal 5-HT1A receptor with WAY-100635. Both the increase and decrease of hypersensitivity induced by amygdaloid glutamate treatment were reversed by medullary administration of a 5-HT1A agonist, 8-OH-DPAT, that presumably produced autoinhibition of serotonergic cell bodies in the medullary raphe. The results indicate that depending on the dose, glutamate in the CeA has a descending facilitatory or inhibitory effect on neuropathic pain hypersensitivity. Serotoninergic raphe neurons are involved in mediating both of these effects. Spinally, the 5-HT3 receptor contributes to the increase and the 5-HT1A receptor to the decrease of neuropathic hypersensitivity induced by amygdaloid glutamate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutaminsäure Mononatriumsalz Hydrat, ≥99% (HPLC), powder
Sigma-Aldrich
Lidocain, powder
Supelco
Lidocain, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutaminsäure Mononatriumsalz Hydrat, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
N-Methyl-D-Asparaginsäure, ≥98% (TLC), solid
USP
Lidocain, United States Pharmacopeia (USP) Reference Standard
USP
Naloxon, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Lidocain, analytical standard
Sigma-Aldrich
Atipamezole, ≥98% (HPLC)
Lidocain, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutaminsäure Mononatriumsalz Hydrat, analytical standard