Merck
  • Insight into conformational modification of alpha-synuclein in the presence of neuronal whole cells and of their isolated membranes.
Insight into conformational modification of alpha-synuclein in the presence of neuronal whole cells and of their isolated membranes.
FEBS letters (2015-02-24)
Giovanni Smaldone, Donatella Diana, Loredano Pollegioni, Sonia Di Gaetano, Roberto Fattorusso, Emilia Pedone
ZUSAMMENFASSUNG
A change in the conformational plasticity of α-Synuclein (α-Syn) is hypothesised to be a key step in the pathogenic mechanism of Parkinson's disease (PD). Here, we report the study of extracellular α-Syn interaction with whole cells and membranes isolated from the neuronal SH-SY5Y cells, exploiting NMR and CD spectroscopies. In addition, the crosslinking agent DSG was used to freeze the conformational and oligomeric state of α-Syn in the presence of cells. These data, in a quasi-physiological environment, confirm the protein monomeric state with a propensity to adopt a transient alpha helical following interaction with biological membranes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Di(N-succinimidyl)-Glutarat, ≥97.0% (CHN)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)