Merck
  • Enhanced nonpeptidergic intraepidermal fiber density and an expanded subset of chloroquine-responsive trigeminal neurons in a mouse model of dry skin itch.
Enhanced nonpeptidergic intraepidermal fiber density and an expanded subset of chloroquine-responsive trigeminal neurons in a mouse model of dry skin itch.
The journal of pain : official journal of the American Pain Society (2015-02-03)
Manouela V Valtcheva, Vijay K Samineni, Judith P Golden, Robert W Gereau, Steve Davidson
ZUSAMMENFASSUNG
Chronic pruritic conditions are often associated with dry skin and loss of epidermal barrier integrity. In this study, repeated application of acetone and ether followed by water (AEW) to the cheek skin of mice produced persistent scratching behavior with no increase in pain-related forelimb wiping, indicating the generation of itch without pain. Cheek skin immunohistochemistry showed a 64.5% increase in total epidermal innervation in AEW-treated mice compared to water-treated controls. This increase was independent of scratching, because mice prevented from scratching by Elizabethan collars showed similar hyperinnervation. To determine the effects of dry skin treatment on specific subsets of peripheral fibers, we examined Ret-positive, calcitonin gene-related peptide (CGRP)-positive, and glial cell line-derived neurotrophic factor family receptor α3 (GFRα3)-positive intraepidermal fiber density. AEW treatment increased Ret-positive fibers but not CGRP-positive or GFRα3-positive fibers, suggesting that a specific subset of nonpeptidergic fibers could contribute to dry skin itch. To test whether trigeminal ganglion neurons innervating the cheek exhibited altered excitability after AEW treatment, primary cultures of retrogradely labeled neurons were examined using whole-cell patch clamp electrophysiology. AEW treatment produced no differences in measures of excitability compared to water-treated controls. In contrast, a significantly higher proportion of trigeminal ganglion neurons was responsive to the nonhistaminergic pruritogen chloroquine after AEW treatment. We conclude that nonpeptidergic, Ret-positive fibers and chloroquine-sensitive neurons may contribute to dry skin pruritus. This study examines the underlying neurobiological mechanisms of persistent dry skin itch. Our results indicate that nonpeptidergic epidermal hyperinnervation and nonhistaminergic pruritic receptors are potential targets for chronic pruritus.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Diethylether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Diethylether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethylether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Diethylether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Supelco
Aceton, analytical standard
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Diethylether
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Diethylether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethylether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
USP
Aceton, United States Pharmacopeia (USP) Reference Standard