Merck
  • Evidence for a specific host-endosymbiont relationship between 'Rickettsia sp. genotype RF2125' and Ctenocephalides felis orientis infesting dogs in India.
Evidence for a specific host-endosymbiont relationship between 'Rickettsia sp. genotype RF2125' and Ctenocephalides felis orientis infesting dogs in India.
Parasites & vectors (2015-04-18)
Sze-Fui Hii, Andrea L Lawrence, Leigh Cuttell, Rebecca Tynas, Puteri Azaziah Megat Abd Rani, Jan Šlapeta, Rebecca J Traub
ZUSAMMENFASSUNG
Fleas of the genus Ctenocephalides serve as vectors for a number of rickettsial zoonoses, including Rickettsia felis. There are currently no published reports of the presence and distribution of R. felis in India, however, the ubiquitous distribution of its vector Ctenocephalides felis, makes it possible that the pathogen is endemic to the region. This study investigates the occurrence of Rickettsia spp. infection in various subspecies of C. felis infesting dogs from urban areas of Mumbai, Delhi and Rajasthan in India. Individual fleas collected off 77 stray dogs from Mumbai, Delhi and Rajasthan were screened for Rickettsia spp. by a conventional PCR targeting the ompB gene. Further genetic characterisation of Rickettsia-positive fleas was carried out using nested PCR and phylogenetic analysis of partial DNA sequences of the gltA and ompA genes. Ctenocephalides spp. were morphologically and genetically identified by PCR targeting a fragment of cox1 gene. Overall, 56/77 fleas (72.7%), including 22/24 (91.7%) from Delhi, 32/44 (72.7%) from Mumbai and 2/9 (22.2%) from Rajasthan were positive for Rickettsia DNA at the ompB gene. Sequences of gltA fragments confirmed the amplification of Rickettsia sp. genotype RF2125. The ompA gene of Rickettsia sp. genotype RF2125 was characterised for the first time and shown 96% identical to R. felis. Three species of Ctenocephalides were identified, with the Ctenocephalides felis orientis being the dominant flea species (69/77; 89.6%) in India, followed by Ctenocephalides felis felis (8/77; 10.4%). High occurrence of Rickettsia sp. genotype RF2125 in C. felis orientis and the absence of R. felis suggests a specific vector-endosymbiont adaptation and coevolution of the Rickettsia felis-like sp. within subspecies of C. felis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Natriumtrichloracetat, 97%
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~0.025 M in H2O