Merck
  • Expression of anger and ill health in two cultures: an examination of inflammation and cardiovascular risk.
Expression of anger and ill health in two cultures: an examination of inflammation and cardiovascular risk.
Psychological science (2015-01-08)
Shinobu Kitayama, Jiyoung Park, Jennifer Morozink Boylan, Yuri Miyamoto, Cynthia S Levine, Hazel Rose Markus, Mayumi Karasawa, Christopher L Coe, Norito Kawakami, Gayle D Love, Carol D Ryff
ZUSAMMENFASSUNG
Expression of anger is associated with biological health risk (BHR) in Western cultures. However, recent evidence documenting culturally divergent functions of the expression of anger suggests that its link with BHR may be moderated by culture. To test this prediction, we examined large probability samples of both Japanese and Americans using multiple measures of BHR, including pro-inflammatory markers (interleukin-6 and C-reactive protein) and indices of cardiovascular malfunction (systolic blood pressure and ratio of total to HDL cholesterol). We found that the link between greater expression of anger and increased BHR was robust for Americans. As predicted, however, this association was diametrically reversed for Japanese, among whom greater expression of anger predicted reduced BHR. These patterns were unique to the expressive facet of anger and remained after we controlled for age, gender, health status, health behaviors, social status, and reported experience of negative emotions. Implications for sociocultural modulation of bio-physiological responses are discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.