Merck
  • Oral administration of paeoniflorin attenuates allergic contact dermatitis by inhibiting dendritic cell migration and Th1 and Th17 differentiation in a mouse model.
Oral administration of paeoniflorin attenuates allergic contact dermatitis by inhibiting dendritic cell migration and Th1 and Th17 differentiation in a mouse model.
International immunopharmacology (2015-03-07)
Dongmei Shi, Xuefeng Li, Dongmei Li, Quanjing Zhao, Yongnian Shen, Hongxia Yan, Hongjun Fu, Hailin Zheng, Guixia Lu, Ying Qiu, Weida Liu
ZUSAMMENFASSUNG
Allergic contact dermatitis (ACD) is a hapten-specific CD4(+) T-cells mediated inflammatory response of the skin. Its pathomechanism involves 2 phases, an induction phase and an elicitation phase. Langerhans cells (LCs) and dendritic cells (DCs) in the skin play key roles in presenting low molecular weight chemicals (haptens) to the lymph nodes. Therefore, inhibition of the migration of LCs or DCs and T-cell proliferation is each expected to control ACD disease. To explore the effectiveness of paeoniflorin (PF) on the migration of LCs and T-cell proliferation in vivo, we establish a murine model of ACD, promoted by repeated exposure to an allergen (specifically 1-Chloro-2,4-dinitrobenzene (DNCB)). Administration of PF inhibits DC migration in this DNCB-induced model in the induction phase. As a result, epidermal LC density in the elicitation phase increased in PF-treated mice when compared to PF-untreated mice. At the same time, PF reduced IFN-γ(+)CD4(+) and IL-17(+)CD4(+) T cells proliferation (but not IL-4(+)CD4(+) T cells proliferation), leading to an attenuated cutaneous inflammatory response. Consistent with this T-cell proliferation profile, secretions of IFN-γ and IL-17 were reduced and IL-10 secretion increased in PF-treated mice, but production of IL-4 and IL-5 remained unchanged in the skin and blood samples. These results suggest that oral administration of PF can treat and prevent ACD effectively through inhibition of DC migration, and thus decrease the capacity of DCs to stimulate Th1 and Th17 cell differentiation and cytokine production.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ammoniumthiocyanat, ACS reagent, ≥97.5%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
SAFC
L-Glutamin
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Supelco
Aceton, analytical standard
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
1-Chlor-2,4-dinitrobenzol, ≥99%
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
1-Chlor-2,4-dinitrobenzol, 97%
Sigma-Aldrich
Ammoniumthiocyanat, crystals
USP
Aceton, United States Pharmacopeia (USP) Reference Standard
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Supelco
Ammoniumthiocyanat, 99.99% trace metals basis
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture