Merck
  • Sex-specific associations of established and emerging cardiac biomarkers with all-cause mortality in older adults: the ActiFE study.
Sex-specific associations of established and emerging cardiac biomarkers with all-cause mortality in older adults: the ActiFE study.
Clinical chemistry (2014-12-17)
Dhayana Dallmeier, Michael Denkinger, Richard Peter, Kilian Rapp, Allan S Jaffe, Wolfgang Koenig, Dietrich Rothenbacher
ZUSAMMENFASSUNG
N-terminal pro B-type natriuretic peptide (NT-proBNP) has strong prognostic value for all-cause mortality in the general population. High-sensitivity assays now allow detection of cardiac troponins even in asymptomatic populations. We examined the association between NT-proBNP, high-sensitivity cardiac troponin T (hs-cTnT), and hs-cTnI and all-cause mortality in older adults. We conducted a longitudinal cohort study [Activity and Function in the Elderly in Ulm (ActiFE Ulm)] including 1506 community-dwelling adults ≥65 years old with NT-proBNP, hs-cTnT, and hs-cTnI measured at baseline. We evaluated the associations between log-transformed biomarker concentrations and 4-year total mortality, accounting for possible confounders, with Cox proportional hazards models. We observed 125 deaths among 1422 participants (median follow-up 4 years). We detected effect modification by sex for all biomarkers (all P values <0.05) expressed as hazard ratio (HR) for death per 1-unit increment of ln(biomarker concentration) in women (n = 618, 37 deaths) compared with men (n = 804, 88 deaths): HR 2.97 (95% CI 2.04-4.33) vs 1.73 (1.40-2.13) for NT-proBNP; 3.67 (2.31-5.81) vs 2.15 (1.61-2.87) for hs-cTnT; and 3.32 (2.13-5.18) vs 1.92 (1.55-2.38) for hs-cTnI. Among 777 participants with undetectable hs-cTnT (<5 ng/L), hs-cTnI remained associated with all-cause mortality in age- and sex-adjusted analysis. NT-proBNP, hs-cTnT, and hs-cTnI were independently associated with all-cause mortality in older adults. The strength of these associations varied between men and women, emphasizing the need for additional sex-specific research among older people.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.