Merck
  • Comparative study of interactions of aliskiren and AT1 receptor antagonists with lipid bilayers.
Comparative study of interactions of aliskiren and AT1 receptor antagonists with lipid bilayers.
Biochimica et biophysica acta (2014-12-24)
A Sadeghpour, M Rappolt, D Ntountaniotis, P Chatzigeorgiou, K Viras, G Megariotis, M G Papadopoulos, E Siapi, G Mali, T Mavromoustakos
ZUSAMMENFASSUNG
The renin-angiotensin-aldosterone system (RAAS) plays a key role in the regulation of blood pressure. Renin is the rate limiting enzyme of the RAAS and aliskiren is a highly potent and selective inhibitor of the human renin. Renin is known to be active both in the circulating blood stream as well as locally, when bound to the (pro)-renin receptor ((P)RR). In this study we have investigated a possible mechanism of action of aliskiren, in which its accumulation in the plasma membrane is considered as an essential step for effective inhibition. Aliskiren's interactions with model membranes (cholesterol rich and poor) have been investigated by applying different complementary techniques: differential scanning calorimetry (DSC), Raman spectroscopy, magic angle spinning (MAS) nuclear magnetic resonance (NMR) spectroscopy and small- and wide-angle X-ray scattering (SAXS and WAXS). In addition, in silico molecular dynamics (MD) calculations were applied for further confirmation of the experimental data. Aliskiren's thermal effects on the pre- and main transition of dipalmitoyl-phosphatidylcholine (DPPC) membranes as well as its topographical position in the bilayer show striking similarities to those of angiotensin II type 1 receptor (AT1R) antagonists. Moreover, at higher cholesterol concentrations aliskiren gets expelled from the membrane just as it has been recently demonstrated for the angiotensin receptor blocker (ARB) losartan. Thus, we propose that both the AT1R and the (P)RR-bound renin active sites can be efficiently blocked by membrane-bound ARBs and aliskiren when cholesterol rich membrane rafts/caveolae are formed in the vicinity of the receptors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.